Anti-LOC55908 Rabbit Polyclonal Antibody

Supplier: Proteintech
23792-1-AP-150UL
10273-132EA 462.47 USD
10273-132
Anti-LOC55908 Rabbit Polyclonal Antibody
Antibodies
The LOC55908 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LOC55908. This antibody recognizes human antigen. The LOC55908 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: LOC55908
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR