Anti-MECOM Rabbit Polyclonal Antibody

Supplier: Proteintech
23201-1-AP-150UL
10273-128EA 462.47 USD
10273-128
Anti-MECOM Rabbit Polyclonal Antibody
Antibodies
The EVI1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human EVI1. This antibody recognizes human antigen. The EVI1 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: MECOM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR