Anti-WFDC3 Rabbit Polyclonal Antibody

Supplier: Proteintech
24917-1-AP-150UL
10273-076EA 462.47 USD
10273-076
Anti-WFDC3 Rabbit Polyclonal Antibody
Antibodies
The WFDC3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human WFDC3. This antibody recognizes human antigen. The WFDC3 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

Type: Primary
Antigen: WFDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR