Anti-LRMP Rabbit Polyclonal Antibody

Supplier: Proteintech
24772-1-AP-150UL
10273-072EA 462.47 USD
10273-072
Anti-LRMP Rabbit Polyclonal Antibody
Antibodies
The LRMP antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LRMP. This antibody recognizes human antigen. The LRMP antibody has been validated for the following applications: ELISA, IHC analysis.

Type: Primary
Antigen: LRMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR