Anti-ARHGAP21 Rabbit Polyclonal Antibody

Supplier: Proteintech
22183-1-AP-150UL
10273-056EA 462.47 USD
10273-056
Anti-ARHGAP21 Rabbit Polyclonal Antibody
Antibodies
The ARHGAP21 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ARHGAP21. This antibody recognizes human, rat antigen. The ARHGAP21 antibody has been validated for the following applications: ELISA, WB, IF, IHC analysis.

Type: Primary
Antigen: ARHGAP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR