Anti-EXD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17313-1-AP-150UL
10273-038EA 462.47 USD
10273-038
Anti-EXD1 Rabbit Polyclonal Antibody
Antibodies
The EXD1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human EXD1. This antibody recognizes human,mouse,rat antigen. The EXD1 antibody has been validated for the following applications: ELISA, WB, IF analysis.

Type: Primary
Antigen: EXD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR