Anti-ACTRT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16992-1-AP-150UL
10273-034EA 462.47 USD
10273-034
Anti-ACTRT2 Rabbit Polyclonal Antibody
Antibodies
The ACTRT2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ACTRT2. This antibody recognizes human,mouse,rat antigen. The ACTRT2 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: ACTRT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR