Anti-RILP Rabbit Polyclonal Antibody

Supplier: Proteintech
13574-1-AP-150UL
10272-994EA 462.47 USD
10272-994
Anti-RILP Rabbit Polyclonal Antibody
Antibodies
The RILP antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human RILP. This antibody recognizes human antigen. The RILP antibody has been validated for the following applications: ELISA, WB, IF analysis.

Type: Primary
Antigen: RILP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR