Gibson Assembly® Cloning Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E5510S
102715-912EA 226 USD
102715-912
Gibson Assembly® Cloning Kit, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
The Gibson Assembly Cloning Kit combines the power of the Gibson Assembly Master Mix with NEB 5-alpha Competent E.coli, enabling successful assembly of multiple DNA fragments, regardless of fragment length or end compatibility and transformation in just under 2 hours.
The Gibson Assembly Cloning Kit has been optimized for the assembly and cloning of up to 6 fragments. It has been rapidly adopted by the synthetic biology community due to its ease-of-use, flexibility and suitability for large DNA constructs.