Anti-RNase H1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20171
102711-214EA 649.66 USD
102711-214
Anti-RNase H1 Rabbit Polyclonal Antibody
Antibodies
The RNase H1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RNase H1. This antibody reacts with human, zebrafish. The RNase H1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RNase H1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR