Anti-Phosphopantothenate-cysteine ligase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19811
102710-494EA 635.51 USD
102710-494
Anti-Phosphopantothenate-cysteine ligase Rabbit Polyclonal Antibody
Antibodies
The Phosphopantothenate-cysteine ligase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Phosphopantothenate-cysteine ligase. This antibody reacts with human. The Phosphopantothenate-cysteine ligase Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Phosphopantothenate-cysteine ligase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR