Anti-DNA Ligase IV Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-16182
102707-422EA 649.66 USD
102707-422
Anti-DNA Ligase IV Rabbit Polyclonal Antibody
Antibodies
The DNA Ligase IV Antibody from Novus Biologicals is a rabbit polyclonal antibody to DNA Ligase IV. This antibody reacts with human, mouse, rat. The DNA Ligase IV Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: DNA Ligase IV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR