Stainless Steel Pipette Cans, Bellco Glass
Supplier: Bellco Glass
1217-11106
102681-542EA 76.58 USD
102681-542
Stainless Steel Pipette Cans, Bellco Glass
Boxes Pipette Sterilization Containers
Heavy gauge rectangular construction for easy stacking and storage.
  • Silicone pads on inside of top and bottom protect pipette tips
  • Cans will accommodate up to 49 Pasteur pipettes (5.75" pipettes)
Can be sterilized by autoclave or dry heat (do not exceed 400 °F).