DiATOME® UltraTrim Trimming Tools, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
UTT-40-R UTT-R UWTT-40-R UWTT-R
102681-530EA 1658.98 USD
102681-530 102681-532 102681-538 102681-540
DiATOME® UltraTrim Trimming Tools, Electron Microscopy Sciences
Knives
Diatome Ultratrim allows quick, easy and accurate trimming at both room and cryo-temperatures.
Due to the extreme sharpness, less mechanical damage is applied to the sample during trimming.