DiATOME® Jumbo Diamond Knives for Histology, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60-HISJ 60-HISJE 60-HISJR 80-HISJ 80-HISJE 80-HISJR
102680-974EA 8394.57 USD
102680-974 102680-976 102680-978 102681-494 102681-496 102681-498
DiATOME® Jumbo Diamond Knives for Histology, Electron Microscopy Sciences
Knives
The Jumbo boat as well as the adhesive (Pattex compact by Henkel) applied to the side of the sample block increase the distinct advantages
They allow easy production of section ribbons (0.5 to 2 µm), no section loss or folding, the same orientation of all sections, easy collection of section ribbons, multiple ribbons on one glass slide, and perfect for Immuno-histo-chemistry.