Omniprobe End Effector Probe Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75981-CU
102681-338PK 1198.5 USD
102681-338
Omniprobe End Effector Probe Tips, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Type 2 Curved End Effector is designed for low attack angle manipulation and lift-out. The shank is curved to permit low angle approach and solid attachment beneath the sample plane without collision with the surrounding matrix. The Straight End Effector for Xtreme Access probe shaft.
Ordering information: Available in Copper (Cu) Curved End Effector. Available in two versions: Copper (Cu) and Molybdenum (Mo).