Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75960-06
102681-284PK 335.45 USD
102681-284
Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Custom tip designed from tungsten. Tip radius is < 0.5 µm with 6° taper angle for maximum lifetime service. Compatible with Short-Cut™ system for direct conversion to TEM grid
Accessories information: Xtreme Access Compatible Probe Point Holder is for use with 1/2" tungsten probe tips. Comes with a storage vessel. The AP250 AutoProbe™ tip holder uses the tungsten probe tip. Compatible with the Short-Cut™ for direct conversion to TEM Grid. Comes with a storage vial and handle.