Tungsten/Nickel Probe Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75960-01
102681-274PK 421 USD
102681-274
Tungsten/Nickel Probe Tips, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Custom probe tip with a nickel tube shank and tungsten tip for use with Omniprobe systems. Tip radius is 0.5 µm with 13° taper angle for maximum lifetime service.