SecureSlip™ Cell Culture Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70462-12 70462-14
100502-726PK 651.17 USD
100502-726 102681-198
SecureSlip™ Cell Culture Coverglass, Electron Microscopy Sciences
Cover Glasses
Convenient, Secure coverglass that will not move during cultivation
The coverglass is affixed to a thin microscopically transparent silicone base which secure to culture vessels by an electrostatic charge, not an adhesive, to prevent movement during cultivation. The silicone also acts as a hydrophobic barrier to isolate liquid reagents on glass. Individual coverglass with silicone backing can be adhered to a petri dish or plate. These can be easily removed one at a time from the dish during culture. Once removed, coverglass can be affixed to a microscope slide by means of the tacky silicone backing for routine processing, or placed cell side down onto a drop of reagent using the silicone backing as a cover to prevent evaporation.