FlowMesh Sheet, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68505-20
102681-178PK 84.98 USD
102681-178
FlowMesh Sheet, Electron Microscopy Sciences
Sheet Protectors
A Permeable Support Mesh for Gels and Membranes
  • Protects - Place FlowMesh above or below a gel or filter to protect from tearing, bending or folding
  • Sturdy - Supports gels during incubation with nucleic acid probes, antibody probes, equilibration buffers, stain, destain solutions, etc.
  • Versatile - Use as a permeable spacer for stacking as well as for staining and destaining procedures
  • Access - Both sides of a container or plastic bag are assured of constant solution access, which improves the uniformity of hybridizations and binding assays.
  • Convenient - FlowMesh is re-usable and easily cut to size