Sensidyne Gilian BDXII Pump, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
65241-12
102681-160EA 634.63 USD
102681-160
Sensidyne Gilian BDXII Pump, Electron Microscopy Sciences
Samplers Air Samplers
The Gilian BDX II Abatement Air Sampler from Sensidyne is a dependable, low cost personal sampling pump.
  • Flow rates – 500 – 3000 cc/min ( 0.50-3 LPM)
  • Rechargeable battery pack system
  • Built-in rotameter & flow adjust
  • Compact, lightweight, quiet
  • Full one year warranty
It is a “no-frills” model designed for use with asbestos and lead sampling. The Gilian BDX II includes a Pump with battery pack, battery charger, tool kit, 36” tubing, clip assembly and manual.