BioCulture™ Pump, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
65224-50
102681-124EA 1750.13 USD
102681-124
BioCulture™ Pump, Electron Microscopy Sciences
Samplers Air Samplers
The BioCulture Calibration Head is A NIST traceable venture calibrator with 1% accuracy that allow users to calibrate the flow of the BioCulture pump themselves without having to send the pump out for service of calibration.
  • Allows flow calibration at flow rates between 30 – 120 LPM
  • Designed for use with impeller fan type pumps like the BioCulture™, which require calibration devices with very little backpressure resistance
Certifications: complies with a certificate of calibration traceable to NIST standards.