Calibration Micrometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60218-1PG 60218-1PGC
102093-154EA 586.94 USD
102093-154 102680-948
Calibration Micrometer, Electron Microscopy Sciences
Micrometers
This ideal standard provides accurate calibration of any eyepiece, reticules, filar micrometers, or electronic measuring instrument
Features a series of ʽH’ shaped fiducial images in a range of sizes from 0.1 mm through 20 mm, useful for calibration range from 1X to 1000X power.