Stage Micrometer Model SM-5, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60210-5PG 60210-5PGC
102680-932EA 850.11 USD
102680-932 102680-934
Stage Micrometer Model SM-5, Electron Microscopy Sciences
Micrometers
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
Ordering information: The Opal and Quartz are offered as a special only and must be requested.
Packaging: All EMS micrometers come in a sturdy wooden box.