MicroHeat Hot Plate, Model MH2, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
55110-MH2
102680-880EA 1451.58 USD
102680-880
MicroHeat Hot Plate, Model MH2, Electron Microscopy Sciences
Heaters Hot Plates
Thermoplastic gluing under stereo microscopes.
  • Heat stabilized
  • Small
  • Ideal for thermoplastic gluing
MicroHeat hot plate is a heat-stabilizes, combined with power supply, designed for embedding samples and for thermoplastic gluing under stereooptical microscope.
Application:
The MicroHeat was developed to make the embedding procedure easier. Because of MicroHeat is small dimensions, it is can be easily placed on the specimen table of the sterooptical microscope, and with the help of the microscope, the gluing faults can be avoided, this result is very important in case of sample requiring bubble-free sticking (e.g. cross section samples for EM). With all of these features, MicroHeat is useful in the fields of material research, geology, semiconductors and optical industry for investigation of multilayer system, semiconductors and high Tc superconductors, diamond, composite materials, ceramics, glasses, rocks and minerals.
Operation:
Model MH 2 is relatively simple to operate. The temperature of the hot plate can be adjusted continuously in a wide range and is kept constant by the built-in electronic. A control circuit maintains the temperature of the heated surface within ±5 °C. The temperature of the heated surface usually reaches the preset temperature within 1 – 3 minutes. However, it is recommended that to use the hot plate at least 5 minutes after the unit was on.
System Configuration:
Main power supply 100 – 240 VAC, 50/60 Hz
MicroHeat unit.