Odyssey® One-Color Protein Molecular Weight Marker, LI-COR®
Supplier: Li-Cor
928-40000
102673-496EA
293.94
USD
102673-496
Odyssey® One-Color Protein Molecular Weight Marker, LI-COR®
Electrophoresis Reagents
Molecular Weight Markers
The Odyssey One-Color Protein MW Marker provides a ladder of convenient, consistent sizes for both visible and 700 channel near-infrared detection.
- Includes ten protein bands from 10 to 250 kDa
- Three reference bands at 25, 50, and 75 kDa are three times as intense as other bands