Anti-IgG Goat Polyclonal Antibody (IRDye® 680RD)

Supplier: Li-Cor
926-68076
102673-420EA 256.09 USD
102673-420
Anti-IgG Goat Polyclonal Antibody (IRDye® 680RD)
Antibodies
The antibody was isolated from antisera by affinity chromatography using antigens coupled to agarose beads. Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule rat IgG, and with the light chains of other rat immunoglobulins. No reactivity was detected against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reactivity with mouse, bovine, horse, human, and rabbit serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot and In-Cell Western™ assay applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, and small animal imaging.

Western blot: 1:15000; In-Cell Western Assay: 1:800

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680RD
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR