Anti-IgG2 beta Goat Polyclonal Antibody (IRDye® 680LT)

Supplier: Li-Cor
926-68052
102673-406EA 389.82 USD
102673-406
Anti-IgG2 beta Goat Polyclonal Antibody (IRDye® 680LT)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using mouse IgG2b covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy chain of mouse IgG2b. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with mouse IgM, IgG1, IgG2a, IgG3, IgA, pooled human sera, and purified human paraproteins. The conjugate has been specifically tested and qualified for Western blot applications. Recommended for: Western blot, proein array, immunohistochemistry, microscopy, 2D gel detection, and tissue section imaging. NOT RECOMMENDED for: small animal imaging, In-Cell Wesern assays, or On-Cell Western assays.

Western blot: 1:20000

Type: Secondary
Antigen: IgG2 beta
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680LT
Epitope:
Host: Goat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR