Odyssey® EMSA (Gel Shift) Reagents, LI-COR
Supplier: Li-Cor
829-07910
102673-212EA 334.31 USD
102673-212
Odyssey® EMSA (Gel Shift) Reagents, LI-COR
Assays Protein Assays
Gel shift assays, or electrophoretic mobility shift assays (EMSA), provide a simple method to study DNA-protein interactions.
  • Can be easily converted to infrared fluorescent assays
  • Conditions are the same as any other EMSA detection method
  • Creates good substrate for protein binding
  • Complete infrared assays in less than two hours
Existing mobility shift assay protocols can be easily converted to infrared fluorescent assays by replacing the existing DNA oligonucleotides with IRDye® infrared dye end-labeled oligonucleotides
A DNA oligonucleotide end-labeled with IRDye 700 infrared dye is a good substrate for protein binding. Using the Odyssey Infrared Imaging System, IRDye infrared dye labeled DNA detection is linear within a 50-fold dilution range from 9.1 fmol to 0.18 fmol. Infrared assays can be completed in less than two hours with no gel transfer or film exposure. The gel doesn’t even have to be removed from the glass plates for imaging. Plus, the gel can be placed back into the electrophoresis unit and run longer, if necessary.
Binding and electrophoresis conditions are the same as any other EMSA detection method. You can easily adapt existing protocols by replacing radiolabeled oligos with IRDye® 700 and IRDye 800 end-labeled oligos.
Ordering information: IRDye 700 infrared dye-labeled EMSA oligonucleotides are supplied as 25 µL of 50 nM double-stranded DNA.