Chicken Alpha 1-Acid Glycoprotein ELISA
Supplier: ALPCO DIAGNOSTICS MS
41-AGPCH-E01
102672-788EA 1256.92 USD
102672-788
Chicken Alpha 1-Acid Glycoprotein ELISA
Assays ELISAs
The chicken alpha 1-acid glycoprotein ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring AGP in the biological fluids of chickens.
AGP is a 52 kDa serine protease inhibitor (serpin) in blood, which protects tissue from enzymes from inflammatory cells, especially elastase. In certain acute phase inflammatory reactions, AGP is elevated in order to limit the damage caused by activated neutrophil granulocytes and their enzyme elastase. Disorders of AGP include AGP deficiency, a hereditary disorder that can lead to severe tissue breakdown during inflammation. This may result in pulmonary emphysema and liver cirrhosis, in severe cases. Genetic variants of AGP do occur.
Caution: For research use only. Not for use in diagnostic procedures.