2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-hydroxy-5,6,7,8-tetrahydroquinazoline

080960-500MG 080960-1G 080960-5G
102672-096EA 387.69 USD
102672-096 102672-098 102672-100
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol

Hazard: Irritant

Formula: C₈H₁₁N₃O
MDL Number: MFCD00665850
CAS Number: 33081-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR