4-(2-Methoxyphenyl)piperazine-1-carboximidamide sulfate

Supplier: Matrix Scientific

080832-500MG 080832-1G 080832-5G
102671-464EA 193.85 USD
102671-464 102671-466 102671-468
4-(2-Methoxyphenyl)piperazine-1-carboximidamide sulfate
4-(2-Methoxyphenyl)piperazine-1-carboximidamide sulfate

Hazard: Irritant

Formula: C₁₂H₂₀N₄O₅S
MW: 332.38 g/mol
MDL Number: MFCD22422000
CAS Number: 1417569-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR