N-[Amino(imino)methyl]pyrrolidine-1-carboximidamide

Supplier: Matrix Scientific

080798-500MG 080798-1G 080798-5G
102671-266EA 258.46 USD
102671-266 102671-268 102671-270
N-[Amino(imino)methyl]pyrrolidine-1-carboximidamide
N-[Amino(imino)methyl]pyrrolidine-1-carboximidamide

Hazard: Irritant

Formula: C₆H₁₃N₅
MDL Number: MFCD15203590
CAS Number: 553-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR