Tetramethyl-9'-ethoxy-5',5'-dimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Supplier: Matrix Scientific
080784-500MG
080784-1G
102671-226EA
161.54
USD
102671-226
102671-228
Tetramethyl-9'-ethoxy-5',5'-dimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Tetramethyl-9'-ethoxy-5',5'-dimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Hazard: Irritant
Formula:
C₂₆H₂₇NO₉S₃ |
MDL Number:
MFCD00521231 CAS Number: 301655-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...