6-Chloro-N,N'-bis(3-chloro-4-methylphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080744-500MG 080744-1G
102671-048EA 193.85 USD
102671-048 102671-050
6-Chloro-N,N'-bis(3-chloro-4-methylphenyl)-1,3,5-triazine-2,4-diamine
6-Chloro-N,N'-bis(3-chloro-4-methylphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₇H₁₄Cl₃N₅
MW: 394.69 g/mol
MDL Number: MFCD22374951
CAS Number: 1379811-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR