2,8-Dimethyl-N-(2-methylphenyl)-1,2,3,4-tetrahydroquinolin-4-amine

Supplier: Matrix Scientific

080573-500MG 080573-1G 080573-5G
102670-228EA 258.46 USD
102670-228 102670-230 102670-232
2,8-Dimethyl-N-(2-methylphenyl)-1,2,3,4-tetrahydroquinolin-4-amine
2,8-Dimethyl-N-(2-methylphenyl)-1,2,3,4-tetrahydroquinolin-4-amine

Hazard: Irritant

Formula: C₁₈H₂₂N₂
MDL Number: MFCD00183925
CAS Number: 54306-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR