N-(6-Ethyl-4-methylquinazolin-2-yl)guanidine
Supplier: Matrix Scientific
080562-500MG
080562-1G
080562-5G
102670-168EA
193.85
USD
102670-168
102670-170
102670-172
N-(6-Ethyl-4-methylquinazolin-2-yl)guanidine
N-(6-Ethyl-4-methylquinazolin-2-yl)guanidine
Hazard: Irritant
Formula:
C₁₂H₁₅N₅ |
MDL Number:
MFCD01564234 CAS Number: 5361-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...