[2-(4-Chloro-3-methylphenoxy)-1-cyclopropylethyl]amine hydrochloride

Supplier: Matrix Scientific

080406-500MG 080406-1G 080406-5G
102669-504EA 516.92 USD
102669-504 102669-506 102669-508
[2-(4-Chloro-3-methylphenoxy)-1-cyclopropylethyl]amine hydrochloride
[2-(4-Chloro-3-methylphenoxy)-1-cyclopropylethyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD22421978
CAS Number: 1417566-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR