3-(3-Methyl-4-nitro-1H-pyrazol-1-yl)propanoic acid

Supplier: Matrix Scientific

080257-1G
102668-700EA 807.69 USD
102668-700
3-(3-Methyl-4-nitro-1H-pyrazol-1-yl)propanoic acid
3-(3-Methyl-4-nitro-1H-pyrazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₇H₉N₃O₄
MW: 199.17 g/mol
MDL Number: MFCD03419614
CAS Number: 956396-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR