[3-(2,4-Dimethoxyphenyl)-1,2,4-oxadiazol-5-yl]acetonitrile

Supplier: Matrix Scientific

077753-500MG
102667-576EA 516.92 USD
102667-576
[3-(2,4-Dimethoxyphenyl)-1,2,4-oxadiazol-5-yl]acetonitrile
[3-(2,4-Dimethoxyphenyl)-1,2,4-oxadiazol-5-yl]acetonitrile

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₃
MW: 245.24 g/mol
MDL Number: MFCD22056541
CAS Number: 1365962-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR