tert-Butyl-6-(aminomethyl)-1H-indole-1-carboxylate hydrochloride

Supplier: Matrix Scientific

077559-500MG 077559-1G 077559-5G
102666-872EA 516.92 USD
102666-872 102666-874 102666-876
tert-Butyl-6-(aminomethyl)-1H-indole-1-carboxylate hydrochloride
tert-Butyl-6-(aminomethyl)-1H-indole-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O₂
MDL Number: MFCD20441458
CAS Number: 1401425-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR