4-Amino-5-(1H-1,2,3-benzotriazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077491-500MG 077491-1G 077491-5G
102666-546EA 387.69 USD
102666-546 102666-548 102666-550
4-Amino-5-(1H-1,2,3-benzotriazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(1H-1,2,3-benzotriazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₉N₇S
MDL Number: MFCD04035853
CAS Number: 122503-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR