4-Hydroxy-5-methylisophthalic acid

Supplier: Matrix Scientific

077332-500MG 077332-1G
102665-650EA 387.69 USD
102665-650 102665-652
4-Hydroxy-5-methylisophthalic acid
4-Hydroxy-5-methylisophthalic acid

Hazard: Irritant

Formula: C₉H₈O₅
MDL Number: MFCD02925539
CAS Number: 4365-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR