6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione

Supplier: Matrix Scientific

077306-1G 077306-5G
102665-522EA 86.15 USD
102665-522 102665-524
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione

Hazard: Irritant

Formula: C₃H₂BrN₃O₂
Melting Pt: 232…234 °C
MDL Number: MFCD00154757
CAS Number: 4956-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR