5-Pyridin-2-yl-1,3,4-oxadiazole-2-thiol

Supplier: Matrix Scientific

077296-500MG 077296-1G 077296-5G
102665-466EA 516.92 USD
102665-466 102665-468 102665-470
5-Pyridin-2-yl-1,3,4-oxadiazole-2-thiol
5-Pyridin-2-yl-1,3,4-oxadiazole-2-thiol

Hazard: Irritant

Formula: C₇H₅N₃OS
MDL Number: MFCD00139078
CAS Number: 3690-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR