2-Butyl-1H-benzimidazole

Supplier: Matrix Scientific

077287-500MG 077287-1G 077287-5G
102665-418EA 258.46 USD
102665-418 102665-420 102665-422
2-Butyl-1H-benzimidazole
2-Butyl-1H-benzimidazole

Hazard: Irritant

Formula: C₁₁H₁₄N₂
MDL Number: MFCD00714456
CAS Number: 5851-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR