N,N,3-Trimethylbutanamide

Supplier: Matrix Scientific

077197-500MG 077197-1G 077197-5G
102664-994EA 161.54 USD
102664-994 102664-996 102664-998
N,N,3-Trimethylbutanamide
N,N,3-Trimethylbutanamide

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD02861246
CAS Number: 5370-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR