2-Bromo-4,5-difluorobenzotrifluoride ≥95%

Supplier: Matrix Scientific

077069-1G
102664-366EA 320 USD
102664-366
2-Bromo-4,5-difluorobenzotrifluoride ≥95%
2-Bromo-4,5-difluorobenzotrifluoride

Hazard: Irritant

Formula: C₇H₂BrF₅
MW: 260.99 g/mol
MDL Number: MFCD11226699
CAS Number: 112290-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR