2,4,6-Trichloropyridine ≥95%

Supplier: Matrix Scientific

075828-5G 075828-25G
102543-206EA 69.23 USD
102543-206 102664-066
2,4,6-Trichloropyridine ≥95%
2,4,6-Trichloropyridine

Hazard: Toxic

Formula: C₅H₂Cl₃N
MW: 182.44
Boiling Pt: 217…218 °C
Melting Pt: 32…34 °C
Flash Pt: >110 °C
MDL Number: MFCD00971928
CAS Number: 16063-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR