2-Methoxy-5-(trifluoromethyl)phenylboronic acid

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-5-(trifluoromethyl)benzeneboronic acid

035469-1G 035469-5G 035469-25G
101839-450EA 36.92 USD
101839-450 101839-452 102663-674
2-Methoxy-5-(trifluoromethyl)phenylboronic acid
2-Methoxy-5-(trifluoromethyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₈H₈BF₃O₃
MDL Number: MFCD04039222
CAS Number: 240139-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR